Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dimethyl-1H-pyrazol-5-ylamine hydrochloride, 97%, Thermo Scientific™

Catalog No. AC297720050
Click to view available options
Quantity:
Packaging:

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS

103068-64-0

Molecular Formula

C5H10ClN3

Molecular Weight (g/mol)

147.606

InChI Key

MOGGYYLZLBLICG-UHFFFAOYSA-N

Synonym

1,3-dimethyl-1h-pyrazol-5-ylamine hydrochloride, 2,5-dimethylpyrazol-3-amine hydrochloride, 5-amino-1,3-dimethylpyrazole hydrochloride, 1,3-dimethyl-1h-pyrazol-5-amine hydrochloride, 1,3-dimethylpyrazole-5-ylamine, chloride, 1h-pyrazol-5-amine, 1,3-dimethyl-, monohydrochloride, c5h9n3.clh, 1h-pyrazol-5-amine, 1,3-dimethyl-, hydrochloride, 1h-pyrazol-5-amine,1,3-dimethyl-,hydrochloride 1:1, 1,3-dimethyl-1h-pyrazol-5-amine-hydrogen chloride 1/1

PubChem CID

2725049

IUPAC Name

2,5-dimethylpyrazol-3-amine;hydrochloride

SMILES

CC1=NN(C(=C1)N)C.Cl

Specifications

CAS Min %

98.5

CAS Max %

100.0

Melting Point

227°C to 230°C

Color

Yellow

Infrared Spectrum

Authentic

Assay Percent Range

99%

Packaging

Glass bottle

Quantity

5 g

Formula Weight

147.61

Percent Purity

≥98.5% (ex Cl)

Physical Form

Crystalline Powder

Chemical Name or Material

5-Amino-1, 3-dimethylpyrazole hydrochloride

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.