Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dimethylurea 98.0+%, TCI America™
SDP

Catalog No. p-7126126 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
500 g

Chemical Identifiers

CAS 96-31-1
Molecular Formula C3H8N2O
Molecular Weight (g/mol) 88.11
MDL Number MFCD00008286
InChI Key MGJKQDOBUOMPEZ-UHFFFAOYSA-N
Synonym n,n'-dimethylurea, sym-dimethylurea, urea, n,n'-dimethyl, dimethylurea, symmetric dimethylurea, urea, 1,3-dimethyl, n,n'-dimethylharnstoff, 1,3-dimethyl urea, n,n'-dimethylharnstoff german, unii-wam6dr9i4x
PubChem CID 7293
ChEBI CHEBI:80472
IUPAC Name 1,3-dimethylurea
SMILES CNC(=O)NC

Specifications

Melting Point 105°C
Color White
Boiling Point 270°C
Quantity 25 g
Formula Weight 88.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dimethylurea
EINECSNumber (2)-1734
RTECSNumber YS9868000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.