Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-[4-(2-Phenylethyl)benzyl]naphthalene 98.0+%, TCI America™
SDP

Catalog No. p-7134992 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
25 g

Chemical Identifiers

CAS 127833-53-8
Molecular Formula C25H22
Molecular Weight (g/mol) 322.451
MDL Number MFCD00191572
InChI Key UKRRCRMJNMWWMZ-UHFFFAOYSA-N
Synonym 1-[4-(2-Phenethyl)benzyl]naphthalene
PubChem CID 14833659
IUPAC Name 1-[[4-(2-phenylethyl)phenyl]methyl]naphthalene
SMILES C1=CC=C(C=C1)CCC2=CC=C(C=C2)CC3=CC=CC4=CC=CC=C43

Specifications

Melting Point 66°C
Color White-Yellow
Quantity 1 g
Formula Weight 322.45
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-[4-(2-Phenylethyl)benzyl]naphthalene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.