Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Benzodioxane-5-carboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. p-7123211 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 4442-53-9
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00239415
InChI Key VCLSWKVAHAJSFL-UHFFFAOYSA-N
Synonym 1,4-benzodioxan-5-carboxylic acid, 2,3-dihydrobenzo b 1,4 dioxine-5-carboxylic acid, 2,3-dihydro-1,4-benzodioxin-5-carboxylic acid, 1,4-benzodioxane-5-carboxylic acid, 2,3-dihydro-benzo 1,4 dioxine-5-carboxylic acid, 2,3-ethylenedioxybenzoic acid, 2,3-dihydro-1,4-benzodioxane-5-carboxylic acid, 2,3-dihydrobenzo 1,4 dioxine-5-carboxylic acid, 1,4-benzodioxin-5-carboxylic acid, 2,3-dihydro, 2h,3h-benzo e 1,4-dioxin-5-carboxylic acid
PubChem CID 78184
IUPAC Name 2,3-dihydro-1,4-benzodioxine-5-carboxylic acid
SMILES C1COC2=C(C=CC=C2O1)C(=O)O

Specifications

Melting Point 192°C
Quantity 5 g
Formula Weight 180.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Benzodioxane-5-carboxylic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.