Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Bis(2-chloroethyl)benzene 96.0+%, TCI America™
SDP

Catalog No. B22011G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 7379-84-2
Molecular Formula C10H12Cl2
Molecular Weight (g/mol) 203.106
InChI Key GGMHFGIZPKYWGQ-UHFFFAOYSA-N
PubChem CID 2749146
IUPAC Name 1,4-bis(2-chloroethyl)benzene
SMILES C1=CC(=CC=C1CCCl)CCCl

Specifications

Melting Point 46°C
Color White-Yellow
Quantity 1 g
Formula Weight 203.11
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Bis(2-chloroethyl)benzene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.