Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dibromo-2-butanol 95.0+%, TCI America™
SDP

Catalog No. p-7126906 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 19398-47-1
Molecular Formula C4H8Br2O
Molecular Weight (g/mol) 231.915
MDL Number MFCD00000252
InChI Key PSSRAPMBSMSACN-UHFFFAOYSA-N
Synonym 1,4-dibromo-2-butanol, 2-butanol, 1,4-dibromo, 2-butanol,1,4-dibromo, 2-butanol,4-dibromo, acmc-1b9jv, 1,4-dibromo-butan-2-ol, wln: e2yq1e, 4-01-00-01581 beilstein handbook reference, racemic, 1 4-dibromo-2-hydroxybutane
PubChem CID 29570
IUPAC Name 1,4-dibromobutan-2-ol
SMILES C(CBr)C(CBr)O

Specifications

Boiling Point 118°C
Quantity 5 g
Formula Weight 231.92
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Dibromo-2-butanol
RTECSNumber EL1515000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.