Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-dimethylpiperazine-2-carboxylic acid dihydrochloride, Thermo Scientific™

Catalog No. p-4391328
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 1174064-55-1
Molecular Formula C7H16Cl2N2O2
Molecular Weight (g/mol) 231.117
InChI Key UPJQXMOMHHDOSZ-UHFFFAOYSA-N
Synonym 1,4-dimethylpiperazine-2-carboxylic acid dihydrochloride, 1,4-dimethylpiperazine-2-carboxylic acid,dihydrochloride, 1,4-dimethylpiperazine-2-carboxylic acid, chloride, chloride, 1,4-dimethylpiperazine-2-carboxylic acid-hydrogen chloride 1/2
PubChem CID 45594293
IUPAC Name 1,4-dimethylpiperazine-2-carboxylic acid;dihydrochloride
SMILES CN1CCN(C(C1)C(=O)O)C.Cl.Cl

Specifications

Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 231.12
Percent Purity 95%
Chemical Name or Material 1, 4-dimethylpiperazine-2-carboxylic acid dihydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.