Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dioxan-2-one 98.0+%, TCI America™
SDP

Catalog No. p-7128384 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 3041-16-5
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.089
MDL Number MFCD00215855
InChI Key VPVXHAANQNHFSF-UHFFFAOYSA-N
Synonym p-Dioxanone
PubChem CID 18233
IUPAC Name 1,4-dioxan-2-one
SMILES C1COC(=O)CO1

Specifications

Melting Point 28°C
Color White
Boiling Point 215°C
Quantity 5 g
Physical Form Crystal-Lump at 20°C
Formula Weight 102.09
Percent Purity ≥98.0% (GC)
Chemical Name or Material 1,4-Dioxan-2-one
RTECSNumber JH2290000
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.