Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Phenylenediamine Sulfate 98.0+%, TCI America™
SDP

Catalog No. p-7134469 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
500 g

Chemical Identifiers

CAS 16245-77-5
Molecular Formula C6H10N2O4S
Molecular Weight (g/mol) 206.22
MDL Number MFCD00035510
InChI Key UFPKLWVNKAMAPE-UHFFFAOYSA-N
Synonym p-phenylenediamine sulfate, benzene-1,4-diamine sulfate, 1,4-diaminobenzene sulfate, 1,4-phenylenediamine sulfate, 1,4-benzenediamine, sulfate 1:1, unii-s7igm30961, benzene-1,4-diamine; sulfuric acid, 1,4-benzenediamine, sulfate, benzene-1,4-diammonium sulphate, 1,4-benzenediamine sulfate
PubChem CID 27769
IUPAC Name benzene-1,4-diamine; sulfuric acid
SMILES OS(O)(=O)=O.NC1=CC=C(N)C=C1

Specifications

Color Purple-White
Quantity 25 g
UN Number 2811
Formula Weight 206.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Phenylenediamine Sulfate
EINECSNumber (3)-0185
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.