Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Piperidyl)-1H-1,2,3-benzotriazole hydrochloride, ≥90%, Thermo Scientific™

Catalog No. p-2694403
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 79098-80-9
Molecular Formula C11H15ClN4
Molecular Weight (g/mol) 238.719
MDL Number MFCD00831110
InChI Key SIZIQJKLJZWLLD-UHFFFAOYSA-N
Synonym 1-4-piperidyl-1h-1,2,3-benzotriazole hydrochloride, 1-piperidin-4-ylbenzotriazole hydrochloride, 1-piperidin-4-yl benzotriazole hydrochloride, 1-piperidin-4-yl-1h-benzotriazole hydrochloride, 1-piperidin-4-yl-1,2,3-benzotriazole hydrochloride, c11h14n4.hcl, 4-piperidylbenzotriazole, chloride, 4-benzotriazol-1-yl-piperidine hcl salt, 4-benzotriazol-1-ylpiperidine hydrochloride, 1-4-piperidinyl benzotriazole hydrochloride
PubChem CID 2779380
IUPAC Name 1-piperidin-4-ylbenzotriazole;hydrochloride
SMILES C1CNCCC1N2C3=CC=CC=C3N=N2.Cl

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 238.72
Percent Purity 90%
Chemical Name or Material 1-(4-Piperidyl)-1H-1, 2, 3-benzotriazole hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful in contact with skin.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.