Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Bis(diphenylphosphino)naphthalene 97.0+%, TCI America™
SDP

Catalog No. p-7123597 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 153725-04-3
Molecular Formula C34H26P2
Molecular Weight (g/mol) 496.53
MDL Number MFCD04117326
InChI Key QOXZKPURCFVBRR-UHFFFAOYSA-N
PubChem CID 2756729
IUPAC Name [8-(diphenylphosphanyl)naphthalen-1-yl]diphenylphosphane
SMILES C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC2=CC=CC(P(C3=CC=CC=C3)C3=CC=CC=C3)=C12

Specifications

Melting Point 210°C
Color Yellow
Quantity 1 g
Formula Weight 496.53
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,8-Bis(diphenylphosphino)naphthalene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.