Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Allyl-3-methylimidazolium Chloride 98.0+%, TCI America™
SDP

Catalog No. p-7117720 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 65039-10-3
Molecular Formula C7H11ClN2
Molecular Weight (g/mol) 158.629
MDL Number MFCD07784450
InChI Key QVRCRKLLQYOIKY-UHFFFAOYSA-M
Synonym 1-allyl-3-methylimidazolium chloride, 1-allyl-3-methyl-1h-imidazol-3-ium chloride, 1-methyl-3-prop-2-en-1-yl imidazol-1-ium chloride, amim-cl, 1-allyl-3-methylimidazoliumchloride, dsstox_cid_27904, dsstox_rid_82657, dsstox_gsid_47928, 1-methyl-3-allylimidazolium chloride, 1-allyl-3-methyl-3-imidazolium chloride
PubChem CID 11321106
IUPAC Name 1-methyl-3-prop-2-enylimidazol-1-ium;chloride
SMILES C[N+]1=CN(C=C1)CC=C.[Cl-]

Specifications

Quantity 5 g
Formula Weight 158.63
Percent Purity ≥98.0% (HPLC,T)
Physical Form Liquid
Chemical Name or Material 1-Allyl-3-methylimidazolium Chloride
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.