Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzhydryl-3-azetidinone 96.0+%, TCI America™
SDP

Catalog No. p-7123757 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 40320-60-3
Molecular Formula C16H15NO
Molecular Weight (g/mol) 237.302
MDL Number MFCD00964539
InChI Key AVUDXLOVIBJFQA-UHFFFAOYSA-N
Synonym 1-diphenylmethyl azetidin-3-one, 1-benzhydryl-azetidin-3-one, 1-1,1-diphenylmethyl azetidin-3-one, 1-diphenylmethyl-3-azetidinone, 1-benzhydryl-3-azetidinone, 3-azetidinone, 1-diphenylmethyl, pubchem10151, acmc-209jd2, n-benzhydrylazetidin-3-one
PubChem CID 3735558
IUPAC Name 1-benzhydrylazetidin-3-one
SMILES C1C(=O)CN1C(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 77°C
Color White-Yellow
Boiling Point 355°C
Quantity 1 g
Formula Weight 237.30
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzhydryl-3-azetidinone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.