Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl-1H-indol-5-ylamine, 97%, Thermo Scientific™

Catalog No. MO00835DA
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 26807-73-8
Molecular Formula C15H14N2
Molecular Weight (g/mol) 222.291
MDL Number MFCD03070173
InChI Key UYDNPZLYDODKKA-UHFFFAOYSA-N
Synonym 1-benzyl-1h-indol-5-ylamine, 1-benzyl-1h-indol-5-amine, 5-amino-1-benzylindole, n-benzyl-5-amino indole, 1-benzylindole-5-ylamine, 5-amino-1benzyl-1h-indole, 5-amino-1-benzyl-1h-indole, 1h-indol-5-amine,1-phenylmethyl
PubChem CID 2794624
IUPAC Name 1-benzylindol-5-amine
SMILES C1=CC=C(C=C1)CN2C=CC3=C2C=CC(=C3)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 222.29
Percent Purity 97%
Chemical Name or Material 1-Benzyl-1H-indol-5-ylamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.