Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzylhydantoin 98.0+%, TCI America™
SDP

Catalog No. B291325G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g

Chemical Identifiers

CAS 6777-05-5
Molecular Formula C10H10N2O2
Molecular Weight (g/mol) 190.20
MDL Number MFCD00466790
InChI Key VJUNTPRQTFDQMF-UHFFFAOYSA-N
PubChem CID 2735485
IUPAC Name 1-benzylimidazolidine-2,4-dione
SMILES C1C(=O)NC(=O)N1CC2=CC=CC=C2

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 190.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzylhydantoin
EINECSNumber (5)-0445
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.