Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-[Bis(trifluoromethanesulfonyl)methyl]-2,3,4,5,6-pentafluorobenzene 98.0+%, TCI America™
SDP

Catalog No. p-7119601 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
100 mg
1 g

Chemical Identifiers

CAS 405074-81-9
Molecular Formula C9HF11O4S2
Molecular Weight (g/mol) 446.205
MDL Number MFCD04117910
InChI Key RLLDXJXYMKTGPV-UHFFFAOYSA-N
Synonym alpha,alpha-Bis(trifluoromethanesulfonyl)-2,3,4,5,6-pentafluorotoluene, 2,3,4,5,6-Pentafluorophenylbis(trifluoromethanesulfonyl)methane
PubChem CID 10203986
IUPAC Name 1-[bis(trifluoromethylsulfonyl)methyl]-2,3,4,5,6-pentafluorobenzene
SMILES C1(=C(C(=C(C(=C1F)F)F)F)F)C(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F

Specifications

Melting Point 89°C
Quantity 100 mg
UN Number 3261
Formula Weight 446.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-[Bis(trifluoromethanesulfonyl)methyl]-2,3,4,5,6-pentafluorobenzene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.