Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-3,3,3-trifluoroacetone 95.0+%, TCI America™
SDP

Catalog No. p-7118943 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 431-35-6
Molecular Formula C3H2BrF3O
Molecular Weight (g/mol) 190.95
MDL Number MFCD00039237
InChI Key ONZQYZKCUHFORE-UHFFFAOYSA-N
Synonym 3-bromo-1,1,1-trifluoroacetone, 1-bromo-3,3,3-trifluoroacetone, bromotrifluoroacetone, 3-bromo-1,1,1-trifluoro-2-propanone, 3-bromo-1,1,1-trifluoropropanone, 1-bromo-3,3,3-trifluoro-2-propanone, 2-propanone, 3-bromo-1,1,1-trifluoro, 1,1,1-trifluoro-3-bromopropanone, 3-bromo-1,1,1-trifluoro-propan-2-one, bromotrifloroacetone
PubChem CID 79008
IUPAC Name 3-bromo-1,1,1-trifluoropropan-2-one
SMILES FC(F)(F)C(=O)CBr

Specifications

Color Yellow
Boiling Point 88°C
Quantity 5 g
UN Number 2924
Formula Weight 190.95
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-3,3,3-trifluoroacetone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.