Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-6-chlorohexane 95.0+%, TCI America™
SDP

Catalog No. p-7119409 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
500 g

Chemical Identifiers

CAS 6294-17-3
Molecular Formula C6H12BrCl
Molecular Weight (g/mol) 199.52
MDL Number MFCD00001019
InChI Key JTYUIAOHIYZBPB-UHFFFAOYSA-N
Synonym 6-chlorohexyl bromide, hexane, 1-bromo-6-chloro, 1-chloro-6-bromohexane, hexyl bromide, 6-chloro, 6-bromohexyl chloride, 1-bromo-6-chloro hexane, 1-bromo-6-chloro-hexane, acmc-1b9m0, 1-bromo-6-chlorohexane, jtyuiaohiyzbpb-uhfffaoysa
PubChem CID 80516
IUPAC Name 1-bromo-6-chlorohexane
SMILES ClCCCCCCBr

Specifications

Color Colorless
Boiling Point 96°C
Quantity 25 g
Formula Weight 199.52
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-6-chlorohexane
EINECSNumber (2)-3316
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.