Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Bromomethyl)-4-phenoxybenzene, ≥97%, Thermo Scientific™

Catalog No. p-3174820
Click to view available options
Quantity:
250 mg
1 g

Chemical Identifiers

CAS 36881-42-2
Molecular Formula C13H11BrO
Molecular Weight (g/mol) 263.134
MDL Number MFCD04038693
InChI Key CPIGBCFBFZSCQI-UHFFFAOYSA-N
PubChem CID 1514251
IUPAC Name 1-(bromomethyl)-4-phenoxybenzene
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)CBr

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Synonym 1-bromomethyl-4-phenoxybenzene, 4-phenoxybenzyl bromide, 1-bromomethyl-4-phenoxy-benzene, benzene,1-bromomethyl-4-phenoxy, 4-phenyloxybenzylbromide, 4-bromomethyl-1-phenoxybenzene, benzene, 1-bromomethyl-4-phenoxy
Formula Weight 263.13
Percent Purity 97%
Chemical Name or Material 1-(Bromomethyl)-4-phenoxybenzene
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.