Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Bromomethyl)isoquinoline hydrobromide, 97%, Thermo Scientific™

Catalog No. p-2694506
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 337508-56-2
Molecular Formula C10H9Br2N
Molecular Weight (g/mol) 302.997
MDL Number MFCD03086096
InChI Key RWSWRJTWPXSENX-UHFFFAOYSA-N
Synonym 1-bromomethyl isoquinoline hydrobromide, 1-bromomethylisoquinoline hydrobromide, isoquinoline, 1-bromomethyl-, hydrobromide 1:1, 1-bromomethylisoquinoline hbr, 1-bromomethyl-isoquinolin hydrobromide, 1-bromomethyl-isoquinoline hydrobromide, isoquinoline, 1-bromomethyl-, hydrobromide
PubChem CID 2776245
IUPAC Name 1-(bromomethyl)isoquinoline;hydrobromide
SMILES C1=CC=C2C(=C1)C=CN=C2CBr.Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 303
Percent Purity 97%
Chemical Name or Material 1-(Bromomethyl)isoquinoline hydrobromide
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.