Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromopentadecane 98.0+%, TCI America™
SDP

Catalog No. p-7118756 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 mL
250 mL

Chemical Identifiers

CAS 629-72-1
Molecular Formula C15H31Br
Molecular Weight (g/mol) 291.32
MDL Number MFCD00000229
InChI Key JKOTZBXSNOGCIF-UHFFFAOYSA-N
Synonym pentadecyl bromide, pentadecane, 1-bromo, 1-bromo pentadecane, n-pentadecyl bromide, pentadecane,1-bromo, 1-bromopentadecane, acmc-209nb8, dsstox_cid_29296, dsstox_rid_83414, dsstox_gsid_49339
PubChem CID 12394
IUPAC Name 1-bromopentadecane
SMILES CCCCCCCCCCCCCCCBr

Specifications

Color Yellow
Boiling Point 158°C
Quantity 25 mL
Formula Weight 291.32
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromopentadecane
EINECSNumber (2)-0067
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.