Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ 1-Butanol
SCI_ED

Catalog No. p-4416499 Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
100 mL
500 mL
4 L
Packaging:
Reagent Grade

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Color Colorless
Density 0.81g/mL
Quantity 100 mL
Packaging Reagent Grade
Assay Percent Range >99%
Linear Formula CH3(CH2)2CH2OH
Formula Weight 74.12
Grade Reagent
Chemical Name or Material 1-Butanol
ShelfLife 2 years
DOTInformation DOT Class 3, III, : Flammable Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.