Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-3-methylimidazolium Dicyanamide 96.0+%, TCI America™
SDP

Catalog No. p-7124077 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 448245-52-1
Molecular Formula C10H15N5
Molecular Weight (g/mol) 205.265
MDL Number MFCD07784457
InChI Key ICIVTHOGIQHZRY-UHFFFAOYSA-N
Synonym 1-butyl-3-methylimidazolium dicyanamide, 1-butyl-3-methylimidazolium dicyamide, 1-butyl-3-methylimidazolium; dicyanoazanide, bmim dca, dsstox_cid_27886, dsstox_rid_82639, dsstox_gsid_47910, c4mim; dicyanoazanide, 1-butyl-3-methyl-1h-imidazol-3-ium cyanoimino methylidene azanide, 1-butyl-3-methylimidazolium; cyanoimino methylidene azanide
PubChem CID 11171745
IUPAC Name 1-butyl-3-methylimidazol-3-ium;cyanoiminomethylideneazanide
SMILES CCCCN1C=C[N+](=C1)C.C(=[N-])=NC#N

Specifications

Melting Point -6°C
Quantity 5 g
Formula Weight 205.27
Percent Purity ≥96.0% (HPLC,N)
Physical Form Liquid
Chemical Name or Material 1-Butyl-3-methylimidazolium Dicyanamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.