Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-3-methylimidazolium Thiocyanate 97.0+%, TCI America™
SDP

Catalog No. p-7123342 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 344790-87-0
Molecular Formula C9H15N3S
Molecular Weight (g/mol) 197.3
MDL Number MFCD06798181
InChI Key SIXHYMZEOJSYQH-UHFFFAOYSA-M
PubChem CID 11424151
IUPAC Name 1-butyl-3-methylimidazol-3-ium;thiocyanate
SMILES CCCCN1C=C[N+](=C1)C.C(#N)[S-]

Specifications

Color Yellow
Quantity 5 g
Formula Weight 197.30
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material 1-Butyl-3-methylimidazolium Thiocyanate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.