Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-3-iodobenzene, 97%, Thermo Scientific™

Catalog No. TL00516EB
Click to view available options
Quantity:

Chemical Identifiers

CAS

625-99-0

Molecular Formula

C6H4ClI

Molecular Weight (g/mol)

238.45

MDL Number

MFCD00001046

InChI Key

JMLWXCJXOYDXRN-UHFFFAOYSA-N

Synonym

3-chloroiodobenzene, m-chloroiodobenzene, benzene, 1-chloro-3-iodo, 3-iodochlorobenzene, 3-chloro-1-iodobenzene, 1-chloro-3-iodo-benzene, m-chlorophenyl iodide, m-iodochlorobenzene, metachloroiodobenzene, 3-chloro-iodobenzene

PubChem CID

12269

IUPAC Name

1-chloro-3-iodobenzene

SMILES

ClC1=CC=CC(I)=C1

Specifications

Density

1.9200g/mL

Assay Percent Range

97%

Linear Formula

ClC6H4I

Quantity

25 g

Formula Weight

238.45

Percent Purity

97%

Chemical Name or Material

1-Chloro-3-iodobenzene

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.