Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chlorononane 98.0+%, TCI America™
SDP

Catalog No. p-7124299 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 mL
500 mL

Chemical Identifiers

CAS 2473-01-0
Molecular Formula C9H19Cl
Molecular Weight (g/mol) 162.701
MDL Number MFCD00001023
InChI Key RKAMCQVGHFRILV-UHFFFAOYSA-N
Synonym nonyl chloride, nonane, 1-chloro, n-nonyl chloride, nonane,1-chloro, tetrapropenyl chloride, 1-propene, tetramer, chlorinated, acmc-209gec, 1-chlorononane, dsstox_cid_29111, dsstox_rid_83330
PubChem CID 17185
IUPAC Name 1-chlorononane
SMILES CCCCCCCCCCl

Specifications

Color Colorless
Boiling Point 204°C
Quantity 25 mL
Formula Weight 162.70
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chlorononane
EINECSNumber (2)-0066&(2)-0068
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.