Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Decyl-3-methylimidazolium Chloride 96.0+%, TCI America™
SDP

Catalog No. p-7128748 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 171058-18-7
Molecular Formula C14H27ClN2
Molecular Weight (g/mol) 258.834
MDL Number MFCD03427605
InChI Key HTZVLLVRJHAJJF-UHFFFAOYSA-M
PubChem CID 2734225
ChEBI CHEBI:61348
IUPAC Name 1-decyl-3-methylimidazol-3-ium;chloride
SMILES CCCCCCCCCCN1C=C[N+](=C1)C.[Cl-]

Specifications

Quantity 5 g
Formula Weight 258.83
Percent Purity ≥96.0% (HPLC)
Physical Form Liquid
Chemical Name or Material 1-Decyl-3-methylimidazolium Chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.