Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Dimethylamino-but-1-en-3-one, 98%, Thermo Scientific Chemicals

Catalog No. p-3736276
Click to view available options
Quantity:
10 mL
100 mL
500 mL
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2802-08-6
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00142611
InChI Key QPWSKIGAQZAJKS-SNAWJCMRSA-N
Synonym e-4-dimethylamino but-3-en-2-one, trans-4-dimethylamino-3-buten-2-one, 4-dimethylamino but-3-en-2-one, 3e-4-dimethylamino but-3-en-2-one, 1-dimethylamino-but-1-en-3-one, 4-dimethylamino-3-buten-2-one, 3e-4-dimethylamino-3-buten-2-one, 3-buten-2-one, 4-dimethylamino-, 3e, 3-buten-2-one, 4-dimethylamino, 3-buten-2-one, 4-dimethylamino-, 3e-9ci
PubChem CID 5369152
IUPAC Name (E)-4-(dimethylamino)but-3-en-2-one
SMILES CC(=O)C=CN(C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Brown to Brown-Red
Density 0.9600g/mL
Boiling Point 101°C to 102°C (5.0 mmHg)
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5570 to 1.56
Linear Formula (CH3)2NCHCHC(O)CH3
Quantity 10 mL
Specific Gravity 0.96
Solubility Information Solubility in water: miscible. Other solubilities: miscible with methanol
Formula Weight 113.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Dimethylamino-but-1-en-3-one
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.