Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Ethylnaphthalene 97.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
10 mL
25 mL
Specifications
Specifications
Color | Colorless |
Boiling Point | 260°C |
Quantity | 10 mL |
Formula Weight | 156.23 |
Percent Purity | ≥97.0% (GC) |
Physical Form | Liquid |
Chemical Name or Material | 1-Ethylnaphthalene |
Safety and Handling
RTECSNumber | QJ6950000 |
TSCA | No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction