Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Fluoroadamantane 98.0+%, TCI America™
SDP

Catalog No. p-7129720 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
250 mg
1 g

Chemical Identifiers

CAS 768-92-3
Molecular Formula C10H15F
Molecular Weight (g/mol) 154.228
InChI Key CPWSNJSGSXXVLD-UHFFFAOYSA-N
Synonym 1-Adamantyl Fluoride
PubChem CID 136608
IUPAC Name 1-fluoroadamantane
SMILES C1C2CC3CC1CC(C2)(C3)F

Specifications

Melting Point 257°C
Color White
Quantity 250 mg
Formula Weight 154.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Fluoroadamantane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.