Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hexyl-3-methylimidazolium Hexafluorophosphate 98.0+%, TCI America™
SDP

Catalog No. p-7130893 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 304680-35-1
Molecular Formula C10H19F6N2P
Molecular Weight (g/mol) 312.24
InChI Key YPWSSSRXUOQNMQ-UHFFFAOYSA-N
Synonym 1-hexyl-3-methylimidazolium hexafluorophosphate, 1-hexyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, 1-n-hexyl-3-methylimidazolium hexafluorophosphate, acmc-209hg2, dsstox_cid_27925, dsstox_rid_82678, dsstox_gsid_47949, ksc222c9l, 1-hexyl-3-methyl-imidazol-3-ium hexafluorophosphate, 1-hex-1-yl-3-methyl-1h-imidazol-3-ium hexafluorophosphate
PubChem CID 2734175
IUPAC Name 1-hexyl-3-methyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
SMILES F[P-](F)(F)(F)(F)F.CCCCCCN1C=C[N+](C)=C1

Specifications

Color Yellow
MDL Number MFCD03093296
Quantity 5 g
Formula Weight 312.24
Percent Purity ≥98.0% (N,T)
Physical Form Liquid
Chemical Name or Material 1-Hexyl-3-methylimidazolium Hexafluorophosphate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.