Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Hexyl-3-methylimidazolium Trifluoromethanesulfonate 98.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
5 g
25 g
Specifications
Specifications
Color | Yellow |
Quantity | 5 g |
Formula Weight | 316.34 |
Percent Purity | ≥98.0% (HPLC,T) |
Physical Form | Liquid |
Chemical Name or Material | 1-Hexyl-3-methylimidazolium Trifluoromethanesulfonate |
Safety and Handling
TSCA | No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction