Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Isopropylnaphthalene 85.0+%, TCI America™
SDP

Catalog No. p-7131497 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 6158-45-8
Molecular Formula C13H14
Molecular Weight (g/mol) 170.255
MDL Number MFCD00061059
InChI Key PMPBFICDXLLSRM-UHFFFAOYSA-N
PubChem CID 22535
IUPAC Name 1-propan-2-ylnaphthalene
SMILES CC(C)C1=CC=CC2=CC=CC=C21

Specifications

Melting Point -16°C
Color Yellow
Boiling Point 268°C
Quantity 1 g
Formula Weight 170.26
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Isopropylnaphthalene
EINECSNumber (4)-0961
TSCA Yes
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.