Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Isopropylpiperazine 98.0+%, TCI America™
SDP

Catalog No. p-7131575 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 4318-42-7
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.219
MDL Number MFCD00167971
InChI Key WHKWMTXTYKVFLK-UHFFFAOYSA-N
Synonym 1-isopropylpiperazine, n-isopropylpiperazine, 1-propan-2-yl piperazine, piperazine, 1-isopropyl, isopropyl piperazine, 1-isopropyl-piperazine, 1-1-methylethyl piperazine, n-isopropyl piperazine, 1-isopropyl piperazine, piperazine, 1-1-methylethyl
PubChem CID 78013
IUPAC Name 1-propan-2-ylpiperazine
SMILES CC(C)N1CCNCC1

Specifications

Color Yellow
Boiling Point 178°C
Quantity 5 g
UN Number 1992
Formula Weight 128.22
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Isopropylpiperazine
RTECSNumber TM0860000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.