Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Methyl-2-pyrrolidone 99.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
25 mL
100 mL
500 mL
Specifications
Specifications
Melting Point | -24°C |
Color | Yellow |
pH | 7.7 - 8 |
Boiling Point | 202°C |
Quantity | 25 mL |
Formula Weight | 99.13 |
Percent Purity | ≥99.0% (GC) |
Physical Form | Liquid |
Chemical Name or Material | 1-Methyl-2-pyrrolidone |
Safety and Handling
EINECSNumber | (5)-0113 |
RTECSNumber | UY5790000 |
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction