Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-3-n-octylimidazolium Tetrafluoroborate 97.0+%, TCI America™
SDP

Catalog No. p-7133395 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 244193-52-0
Molecular Formula C12H23BF4N2
Molecular Weight (g/mol) 282.13
MDL Number MFCD03095434
InChI Key GXZCAMSPWNHTAE-UHFFFAOYSA-N
Synonym 1-methyl-3-octylimidazolium tetrafluoroborate, 1-methyl-3-n-octylimidazolium tetrafluoroborate, 3-methyl-1-octylimidazolium tetrafluoroborate, 1-octyl-3-methylimidazolium tetrafluoroborate, omimbf4, dsstox_cid_27919, dsstox_rid_82672, dsstox_gsid_47943, 3-methyl-1-octylimidazoliumtetrafluoroborate, 1-methyl-3-octylimidazolium tetrafluoroborate hplc
PubChem CID 11460221
IUPAC Name 3-methyl-1-octyl-1H-imidazol-3-ium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.CCCCCCCCN1C=C[N+](C)=C1

Specifications

Quantity 5 g
Formula Weight 282.13
Percent Purity ≥97.0% (HPLC,N)
Physical Form Liquid
Chemical Name or Material 1-Methyl-3-n-octylimidazolium Tetrafluoroborate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.