Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1-methyl-5-phenyl-1H-pyrazol-4-yl)methanol, Thermo Scientific™

Catalog No. p-4391417
Click to view available options
Quantity:
250 mg
1 g
5 g

Chemical Identifiers

CAS 499785-47-6
Molecular Formula C11H12N2O
Molecular Weight (g/mol) 188.23
MDL Number MFCD04115390
InChI Key ZVGZXURABHDVPS-UHFFFAOYSA-N
Synonym 1-methyl-5-phenyl-1h-pyrazol-4-yl methanol, 1-methyl-5-phenylpyrazol-4-yl methanol, 1-methyl-5-phenylpyrazol-4-yl methan-1-ol, bck
PubChem CID 2795476
SMILES CN1N=CC(CO)=C1C1=CC=CC=C1

Specifications

Quantity 250 mg
IUPAC Name (1-methyl-5-phenyl-1H-pyrazol-4-yl)methanol
Formula Weight 188.23
Percent Purity 97%
Chemical Name or Material (1-methyl-5-phenyl-1H-pyrazol-4-yl)methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.