Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylxanthine 97.0+%, TCI America™
SDP

Catalog No. p-7133218 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
50 mg
200 mg

Chemical Identifiers

CAS 6136-37-4
Molecular Formula C6H6N4O2
Molecular Weight (g/mol) 166.14
MDL Number MFCD00005561
InChI Key MVOYJPOZRLFTCP-UHFFFAOYSA-N
PubChem CID 80220
ChEBI CHEBI:68444
IUPAC Name 1-methyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
SMILES CN1C(=O)NC2=C(NC=N2)C1=O

Specifications

Quantity 50 mg
Formula Weight 166.14
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methylxanthine
RTECSNumber ZD8575000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.