Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Nitropropane 98.0+%, TCI America™
SDP

Catalog No. p-7133701 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
500 g

Chemical Identifiers

CAS 108-03-2
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.094
MDL Number MFCD00007407
InChI Key JSZOAYXJRCEYSX-UHFFFAOYSA-N
Synonym propane, 1-nitro, nitropropane, propane, nitro, ccris 1329, dsstox_cid_980, dsstox_rid_75902, dsstox_gsid_20980, nitro-propane, n-nitropropane, 3-nitropropane
PubChem CID 7903
ChEBI CHEBI:76261
IUPAC Name 1-nitropropane
SMILES CCC[N+](=O)[O-]

Specifications

Color Yellow
Boiling Point 131°C
Quantity 25 g
UN Number 2608
Formula Weight 89.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Nitropropane
EINECSNumber (2)-3738
RTECSNumber TZ5075000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.