Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Oxa-6-azaspiro[3.4]octane oxalate, 96%, Thermo Scientific Chemicals

Catalog No. p-7038479
Click to view available options
Quantity:
250 mg
500 mg

1-Oxa-6-azaspiro[3.4]octane oxalate is used as a pharmaceutical intermediate in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Oxa-6-azaspiro[3.4]octane oxalate is used as a pharmaceutical intermediate in synthetic chemistry.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 1408074-51-0
Molecular Formula C8H13NO5
Molecular Weight (g/mol) 203.19
MDL Number MFCD19982774
InChI Key ZKTJFOHJBNDMDS-UHFFFAOYNA-N
Synonym 1-oxa-6-azaspiro 3.4 octane oxalate, 1-oxa-6-azaspiro 3.4 octane; oxalic acid
PubChem CID 71432930
IUPAC Name 1-oxa-7-azaspiro[3.4]octane;oxalic acid
SMILES OC(=O)C(O)=O.C1CC2(CCNC2)O1

Specifications

Quantity 250 mg
Formula Weight 203.19
Percent Purity 96%
Chemical Name or Material 1-Oxa-6-azaspiro[3.4]octane oxalate
Hazard Category H302+H312
Hazard Statement GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.