Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-piperidin-4-yl-5-(trifluoromethyl)-1H-1,2,3-benzotriazole hydrochloride, 97%, Thermo Scientific™

Catalog No. KM10214DA
Click to view available options
:

Specifications

CAS

306935-37-5

Assay Percent Range

97%

Molecular Formula

C12H14ClF3N4

Quantity

1g

Synonym

1-piperidin-4-yl-5-trifluoromethyl-1h-1,2,3-benzotriazole hydrochloride, 4-5-trifluoromethyl benzotriazol-1-yl piperidine hydrochloride, 1-piperidin-4-yl-5-trifluoromethyl-1,2,3-benzotriazole hydrochloride, 1-4-piperidyl-5-trifluoromethyl benzotriazole, chloride, 1-piperidin-4-yl-5-trifluoromethyl benzotriazole hydrochloride, 1-piperidin-4-yl-5-trifluoromethyl-1h-benzo d 1,2,3 triazole hydrochloride, 1-piperidin-4-yl-5-trifluoromethyl-1h-benzotriazole-hydrogen chloride 1/1

InChI Key

OBGFRHXXUYUYOS-UHFFFAOYSA-N

SMILES

C1CNCCC1N2C3=C(C=C(C=C3)C(F)(F)F)N=N2.Cl

IUPAC Name

1-piperidin-4-yl-5-(trifluoromethyl)benzotriazole;hydrochloride

Molecular Weight (g/mol)

306.717

PubChem CID

2780683

Formula Weight

333.75

Percent Purity

97%

Chemical Name or Material

1-Piperidin-4-yl-5-(CF3)-1H-1, 2, 3-benzotriazole hy drochloride hydrate

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.