Learn More
1-Propanol, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | -127.0°C |
Density | 0.8000g/mL |
Boiling Point | 97.0°C |
Quantity | 500 mL |
Flash Point | 15°C |
Refractive Index | 1.3840 to 1.3860 |
Specific Gravity | 0.8 |
Linear Formula | CH3CH2CH2OH |
Viscosity | 2.2 mPa.s (20°C) |
Water | 0.2% max. |
Show More |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS H Statement May cause drowsiness or dizziness. Causes serious eye damage. Highly flammable liquid and vapor. |
Precautionary Statement | GHS P Statement Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.