Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

10-DEBC hydrochloride, Tocris Bioscience™

Catalog No. p-4783677 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective Akt/PKB inhibitor

Selective inhibitor of Akt/PKB. Inhibits IGF-1-stimulated phosphorylation and activation of Akt (complete inhibition at 2.5 μM), suppressing downstream activation of mTOR, p70 S6 kinase and S6 ribosomal protein. Shows no activity at PDK1, SGK1 or PI 3-kinase. Inhibits cell growth (IC50 ∽ 2-6 μM) and induces apoptosis in rhabdomyosarcoma cells.

Chemical Identifiers

CAS 925681-41-0
Molecular Formula C20H26Cl2N2O
Molecular Weight (g/mol) 381.341
InChI Key SVKSJUIYYCQZEC-UHFFFAOYSA-N
Synonym Akt Inhibitor X, 10-4′-n-diethylamino butyl-2-chlorophenoxazine, hcl, 10-4'-n,n-diethylamino butyl-2-chlorophenoxazine hydrochloride, 4-2-chlorophenoxazin-10-yl butyl diethylamine hydrochloride, 4-2-chloro-10h-phenoxazin-10-yl-n,n-diethylbutan-1-amine-hydrogen chloride 1/1
PubChem CID 16760284
IUPAC Name 4-(2-chlorophenoxazin-10-yl)-N,N-diethylbutan-1-amine;hydrochloride
SMILES CCN(CC)CCCCN1C2=CC=CC=C2OC3=C1C=C(C=C3)Cl.Cl

Specifications

Quantity 10 mg
Formula Weight 381.34
Percent Purity >99%
Chemical Name or Material 10-DEBC hydrochloride
Recommended Storage Desiccate at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.