Learn More
Ace Glass, Inc. 10 x 20mm porosity A stick GLASSBLOWER
Supplier:  Ace Glass, Inc. 1533703
Encompass_Preferred
10mm O.D. x 20mm long porous fritted (145-174 micron) glass filter stick, used to fabricate gas disperstion tubes. Hollow at one end. Pack of 6.
Catalog No. 50-983-564
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.