Learn More
Inorganic Ventures 100ug mL POTASSIUM 500mL (Inorganic Ventures)
SDP
Supplier:  Inorganic Ventures MSK100PPM500ML
Encompass_Preferred
Inorganic Ventures' 100 ppm (g/mL) Potassium for ICP-MS is a 500mL certified reference material set in a nitric acid matrix for stability. This potassium standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. One year Expiration Date.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. NC1079550
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.