Learn More
Bioassay Systems QuantiFluo™ Cholesterol Uptake Assay Kit. Cholesterol uptake by adherent cells, screening of cholesterol uptake inhibitors, and evaluation of effect of drugs on cholesterol uptake.
SDP
Supplier:  Bioassay Systems DCUT100
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
For quantitatve determination of cholesterol uptake in cells and screening of modulators on cholesterol uptake. Key Features: Convenient. Treat cells directly in 96-well fluorescent plate. Safe. Non-radioactive assay. High-throughput. Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day Method: FL485/535nm. Samples: Adherent cells. Species: All. Procedure: Assay takes 24-72 hrs, hand-on time 1 hr. Kit size: 100 tests.
Catalog No. 50-195-6284