Learn More
Bioassay Systems QuantiChrom™ Β-N-Acetylglucosaminidase Assay Kit.Β-N-Acetylglucosaminidase activity determination in biological samples.
SDP
Supplier:  Bioassay Systems DNAG100
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
For quantitatve determination of β-N-Acetylglucosaminidase activity and evaluation of drug modulators. Key Features: Fast and sensitive. Linear detection range (20 µL sample): 0.2 to 50 U/L for a 30 minute reaction at 37°C. High-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD405nm. Samples: Urine, serum, plasma, cell lysate, etc. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 0.2 U/L.
Catalog No. 50-195-6292