Learn More
Bioassay Systems QuantiChrom™ Glucose Dehydrogenase Assay Kit. For quantitative determination of D-glucose dehydrogenase enzyme activity and drug effects on its metabolism.
SDP
Supplier:  Bioassay Systems DGDH100
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
For quantitative determination of D-glucose dehydrogenase enzyme activity and drug effects on its metabolism. Key Features: Fast and sensitive. Linear detection range (20 µL sample): 0.5 to 200 U/L for 15 min reaction. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD565nm. Samples: Biological samples (e.g. plasma, serum, tissue and culture media). Species: All. Procedure: Assay takes 15 min. Kit size: 100 tests. Detection limit: 0.5 U/L.
Catalog No. 50-195-6287
missing translation for 'QuantityLeft'