Learn More
Bioassay Systems EnzyChrom™ Coenzyme A Assay Kit. For quantitative determination of coenzyme A (CoA) and evaluation of drug effects on CoA metabolism.
Supplier: Bioassay Systems ECOA100
For quantitative determination of coenzyme A (CoA) and evaluation of drug effects on CoA metabolism. Key Features: Sensitive. Use 10 µL samples. Linear detection range: colorimetric assay 5 - 1000 µM, fluorimetric assay 3 - 100 µM CoA. Convenient. Room temperature "mix-and-read" procedure can be readily automated for high-throughput assay of thousands of samples per day. Method: OD570nm, or FL530/585nm. Samples: Biological. Species: All. Procedure: Assay takes 60 min. Kit size: 100 tests. Detection limit: 3 µM.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.