Learn More
Bioassay Systems EnzyChrom™ L-Amino Acid Assay Kit. Quantitative determination of L-Amino Acids in biological and other samples.
SDP
Supplier:  Bioassay Systems ELAA100
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
For quantitative determination of L-Amino Acids in biological and other samples. Key Features: Fast and sensitive. Linear detection range: 3.3 to 500 µM (colorimetric assay) and 0.13 to 50 µM (fluorimetric assay) for 60 min reaction. Convenient. The procedure involves adding a single working reagent and reading after 60 minutes. High-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated to process thousands of samples per day. Method: OD570nm or FL530/585nm. Samples: Serum, culture media, tissue homogenates, cell lysates, urine, food samples, etc. Species: All. Procedure: Assay takes 60 min. Kit size: 100 tests. Detection limit: 3.3 µM (colorimetric assay), 0.13 µM (fluorimetric assay).
Catalog No. 50-195-6315
missing translation for 'QuantityLeft'